EP0030127870
Eppendorf® storage film, self-adhesive
self-adhesive, pkg of 100 ea, PCR clean
Sinonimo/i:
Eppendorf® Microplate Sealing Films & Foils
About This Item
Codice UNSPSC:
45131506
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
PCR clean
self-adhesive
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Eppendorf® 0030127870
Descrizione generale
This self-adhesive storage film is made for use with Eppendorf Standard and Deepwell 96- and 38-Well plates (polypropylene).
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)