Documenti fondamentali
CLS9099
Corning® aspiration pipettes
volume 5 mL
Sinonimo/i:
Aspiration Pipette
About This Item
Codice UNSPSC:
41121500
eCl@ss:
32070201
NACRES:
NB.01
Materiali
polystyrene
Sterilità
sterile
Confezionamento
case of 200 ea
pack of 50 ea
Produttore/marchio commerciale
Corning 9099
Capacità
5 mL
Volume
5 mL
Descrizione generale
  • Corning® aspirating pipettes are made of optically clearpolystyrene for durability and good mechanical strength.
  • The sterile,ungraduated, and unplugged pipettes are best suited for aspirating liquidaseptically using vacuum suction.
  • Individually wrappedand bulk packed.
  • Disposable andcertified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)