Documenti fondamentali
CLS7016560
Corning® VISTA Petri Dish
dish diam. 60 mm × 15 mm
Sinonimo/i:
Filter crucible
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.43
Materiali
round
soda-lime glass dishes with lids
Sterilità
non-sterile
Caratteristiche
lid
reusable
Confezionamento
case of 24
Produttore/marchio commerciale
Corning 70165-60
Diam. piastra
60 mm × 15 mm
Descrizione generale
The thermal expansion of Soda Lime Glass is 93.5 x 10-7 cm/cm/°C. The normal service temperature is 110°C and the extreme service temperature is 460°C. These dishes are not designed for rapid thermal shock. The Corning® brand Petri Dishes should be viewed as a low cost alternative to Pyrex® brand Petri Dishes (Catalog CLS3160) which are made from low expansion (32.5 x 10-7 cm/cm/°C) borosilicate glass.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4