CLS70075100
Pyrex® VISTA™ graduated cylinder, to contain, economy
volume 100 mL
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41121805
NACRES:
NB.43
Prodotti consigliati
Produttore/marchio commerciale
Corning 70075-100
Volume
100 mL
Categorie correlate
Descrizione generale
These Pyrex VISTA cylinders are calibrated "to contain" (TC). They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Tolerances are +/- 5% of total at any point. By removing the base, the graduate can be conveniently stacked. Bumper guards are supplied with 25 mL through 100 mL inclusive. The 10 mL has a funnel top for careful pouring.
Note legali
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS70075100-12EA | 4061831702972 |