CLS4930
Costar® vacuum aspirator
hand-held vacuum aspirator
Sinonimo/i:
Vacuum Aspiration Device
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.11
Prodotti consigliati
Confezionamento
pack of 1
Produttore/marchio commerciale
Corning 4930
Descrizione generale
Hand-held vacuum aspiration device with a wide variety of accessories available for removal of supernatants. The device attaches to your laboratory vacuum supply via a tubulation fitting. Fingertip control of vacuum reduces the risk of losing valuable samples.
Use with pipette tips, Pasteur pipettes, or any of the aspirator accessories below. Unit comes with single-channel adapter for disposable pipette tips.
Use with pipette tips, Pasteur pipettes, or any of the aspirator accessories below. Unit comes with single-channel adapter for disposable pipette tips.
Note legali
Costar is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)