Documenti fondamentali
CLS480125
Corning® LSE single block, for Corning LSE single block digital dry baths
single block, 12 x 16 mm tubes, 1/cs
Sinonimo/i:
equipment replacement accessory
About This Item
Codice UNSPSC:
41102404
NACRES:
NB.15
Caratteristiche
Holds 12 x 16 mm tubes
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
Corning 480125
Compatibilità
suitable for (use with Corning LSE digital dry bath heater, single block)
Descrizione generale
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
Note legali
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)