CLS451536L
Corning® direct overhead drive assembly for ProCulture® spinner flasks
paddle assembly for 36L Proculture spinner flask w/ direct drive overhead motor, 1/cs
Sinonimo/i:
Corning container
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.43
Prodotti consigliati
Sterilità
non-sterile
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
Corning 4515-36L
Descrizione generale
Direct drive paddle assembly for use with direct drive overhead motors on series 4510 and 4512, 36L Proculture® Spinner Flasks. The paddle assemblies can be used with either 100 mm or 120 mm direct drive center cap assemblies.
Note legali
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)