CLS4080
Corning® Lambda® plus 8-channel pipettor
volume 0.5-10 μL
Sinonimo/i:
multi-channel pipettor
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Caratteristiche
8-channel
Produttore/marchio commerciale
Corning 4080
Campo di misura del volume
0.5-10 μL
Volume
10 μL
Descrizione generale
- Corning® Lambda® plus 8-channel pipettor is designed for universal fit with all common brands of pipet tips.
- Factory calibrated to strict ISO 8655 standards-3-year warranty.
- Smooth plunger movement and extremely low pipetting forces.
- Fully autoclavable and UV resistant.
- Adjustable tip ejector.
- Engineered to provide the highest levels of accuracy, reproducibility, and comfort.
- Inclusive of key for easy in-house calibration.
Note legali
Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)