Documenti fondamentali
CLS1300P500W
Corning® wide mouth wash bottle, reusable
capacity 500 mL, low-density polyethylene (LDPE), with GL-45 PP screw cap
Sinonimo/i:
laboratory wash bottles, plastic wash bottles
About This Item
Codice UNSPSC:
41103209
NACRES:
NB.43
Materiali
cylindrical bottle
low-density polyethylene (LDPE)
screw top cap
white bottle
wide-mouth bottle
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Corning 1300P-500W
Alt. × diam. est.
146 mm × 58 mm
Capacità
500 mL
Descrizione generale
  • Corning® reusable bottles are manufactured from low-density polyethylene (LDPE).
  • Features a wide mouth for easy transferring of samples.
  • Polypropylene cap and dispensing tube optimize backflow of the solution.
  • Unique design allows minimal turbulence of liquid in the bottle.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)