C2798
Biohazard incinerator carton
Floor model
Sinonimo/i:
incineration waste container
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Prodotti consigliati
Produttore/marchio commerciale
Bel-Art F13205-0001
Misura
12 in. × 12 in. × 27 in.
Descrizione generale
Designed to be incinerated along with its contents when full for safer disposal of biohazardous material. The sturdy corrugated cardboard box is supplied with a 1.5 mil HMHD liner to contain liquid waste and is imprinted with the "biohazard" pictogram and instructions in four languages.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)