Documenti fondamentali
AXYPCRAS200
Aluminum sealing film for thermal and general sealing
size 35 μm, pkg of 5x100sheets/cs
Sinonimo/i:
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.22
Materiali
aluminum
Sterilità
non-sterile
Caratteristiche
piercable
Produttore/marchio commerciale
Axygen Inc PCR-AS-200
Corning
Confezionamento
pkg of 5x100sheets/cs
Misura
35 μm
Descrizione generale
  • Aluminum sealing films are ideal for light-sensitive samples.
  • Uniform adhesive enables efficient well sealing.
  • Versatile sealing film can be used in various applications including PCR, Real-Time PCR to ELISA and cell culture.
  • Compatible with a wide range of reagents and temperatures.
  • Axygen® aluminum sealing films are ideal for light-sensitive samples.
  • Uniform adhesive enables efficient well sealing.
  • Versatile sealing film can be used in various applications including PCR, Real-Time PCR to ELISA and cell culture.
  • Compatible with a wide range of reagents and temperatures.
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
AXYPCRAS200