AXYPCR384M2G
PCR microplate for ABI 3730, 3100 and MJ Thermocyclers
size 384 wells, pkg of 5x10plates/cs
Sinonimo/i:
384 multiwell plate
About This Item
Materiali
conical bottom green polypropylene wells
Sterilità
non-sterile
Produttore/marchio commerciale
Axygen Inc PCR-384M2-G
Corning
Confezionamento
pkg of 5x10plates/cs
Volume massimo
50 μL
Misura
384 wells
Volume d’esercizio
50 μL
Tipo di binding
non-treated surface