Key Documents
AXYAM24SQ
Corning® Axygen® AxyMats® 24 Well Sealing Mat for Deep Well Plates
for deep well plates, piercable, 24 well
Sinonimo/i:
Sealing mat
About This Item
Codice UNSPSC:
41122410
Materiali
silicone (research-grade)
Sterilità
non-sterile
Caratteristiche
piercable
Produttore/marchio commerciale
Axygen Inc
Corning
Note legali
AxyMats is a registered trademark of Corning, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.