Documenti fondamentali
ADAPF346382-EP
Adapter for Eppendorf® F-34-6-38 Rotor
holds 3 round-bottom tubes 2.6-7 mL, pack of 2 ea
About This Item
Codice UNSPSC:
41103911
Caratteristiche
Holds 2.6 - 7 mL tubes
holds 3 round-bottom tubes 2.6-7 mL
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 022637279
tecniche
centrifugation: suitable
Compatibilità
for use with Rotor F-34-6-38
Note legali
Eppendorf is a registered trademark of Eppendorf AG