Documenti fondamentali
NA18/22
Millipore
NA18 Diaphragm EPDM
NovAseptic Valve, Diaphragm, EPDM
About This Item
Codice UNSPSC:
31411800
Materiali
EPDM rubber device (ethylene propylene diene monomer rubber)
PTFE
Livello qualitativo
Nome Commerciale
NovAseptic®
Caratteristiche
hydrophobic (Surface)
Parametri
-1-(-6) bar pressure range
-30-140 °C temp. range (Short term use)
Peso
0.02 kg (0.044 lb)
Applicazioni
The applied valve body and actuator may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Confezionamento
The Diaphragm is packaged in a closed box.
Altre note
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.