Documenti fondamentali
Z764523
2mag bioMIXcontrol S control unit
AC/DC input 230 V AC, Schuko plug
Sinonimo/i:
induction stirrer, magnetic induction stir plate, magnetic wear-free stirrer, stirrer
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Materiali
stainless steel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
2mag 98500
Parametri
5-250 rpm speed
Largh. × prof. × alt.
215 mm × 120 mm × 55 mm
Dimensioni della piastra × Largh. × Lungh.
× 200 mm × 250 mm
Peso
1.4 kg
Uscita
4-40 W (stirring power)
Descrizione generale
  • Efficient control unit, for 2mag bioMIXdrive series, developed for the hard daily lab use and uncompromising warming-free continuous operation
  • Control unit for the connection of 1 stirring drive bioMIXdrive 1, 2, 3, 4, or 6 with 6 watts/stirring point, without any own-generated heating effect, extended speed range from 5 to 250 rpm, jerk-free stirring, even and smooth stirring also at slow speeds
  • Power setting (10-step) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation, e.g. in CO2-incubators
  • Identical to bioMIXcontrol, but with stackable housing and vertical control panel
  • Digital display for speed- and power setting, SoftStart for reliable and safe accelerating for stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, and easy to clean
Richiesto ma non fornito
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)