Z758973
Bollé Atom safety goggles
ANSI compliant
Sinonimo/i:
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
About This Item
Codice UNSPSC:
46181802
Prodotti consigliati
Caratteristiche
ANSI compliant
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bollé 40092
Descrizione generale
These Bollé Atom safety goggles are lightweight and compact and fit over prescription spectacles as well as with a respiratory half mask. There is an adjustable strap and a liquid overflow chute. The goggles are Thermo Plastic Rubber (TPR) ventilated.
- Anti-scratch, and anti-fog coatings
- Adjustable strap
- Ball-pivot
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)