Documenti fondamentali
Z758868
Bollé strap conversion kit for Cobra safety spectacles
for CE compliant Cobra spectacles
Sinonimo/i:
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
About This Item
Codice UNSPSC:
46181802
NACRES:
NB.35
Caratteristiche
for CE compliant Cobra spectacles
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bollé KITFSCOB
Disponibilità
not available in USA
Descrizione generale
These optional strap conversion kits for the Cobra safety spectacles allow the ability to interchange the temples with an adjustable strap and foam option. Use these kits with the Cobra wrap around safety spectacles, sold separately: Z758833 or Z758841
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)