Z742264
DURAN® GL 56 Bottle Tag
yellow silicone
Sinonimo/i:
292435634
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
yellow silicone
Confezionamento
pack of 20 ea
Produttore/marchio commerciale
DURAN® 292435634
Categorie correlate
Descrizione generale
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Caratteristiche e vantaggi
- Manufactured from silicone rubber
- The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
- The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and reusable
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742264-20EA | 4061832802404 |