Documenti fondamentali
Z742018
Duran® Bottle Tags GL 45
Eight colors: two bottle tags are supplied with each color
Sinonimo/i:
292432904
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Confezionamento
pack of 16
Produttore/marchio commerciale
DURAN® 292432904
Descrizione generale
  • Colourful silicone GL 45 Bottle Tags can be easily and securely attached around the neck of the YOUTILITY bottle
  • Distinct colours for easy customisation and unambiguous bottle identification
  • Elastic bottle tags will fit around any DIN GL 45 bottle neck
  • Can be applied with the screw cap in place
  • From silicone, eight colours, pack of 16
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)