Z742010
Duran® Youtility bottle, GL 45, complete
clear glass, capacity 1000 mL
Sinonimo/i:
Duran® Youtility
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
clear glass
polypropylene cap
round bottle
Giunto
Joints threaded neck (GL45)
Sterilità
non-sterile
Caratteristiche
graduations
with cap
Confezionamento
pack of 4 ea
Produttore/marchio commerciale
DURAN® 218815457
Capacità
1000 mL
Categorie correlate
Descrizione generale
- Specially shaped gripping zones on both sides of the bottle for easier and safer handling
- Opening or closing of the bottle is faster, and fully compatible with DIN GL 45 closures and other accessories
- Slimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
- Pre-defined labelling area is compatible with DURAN self-adhesive YOUTILITY bottle labels
- Nominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumes
- Sequentially sized bottles are available in four volumes
- Moulded from the DURAN borosilicate 3.3 pharmacopeia type 1 neutral glass
- Retrace Code for full product traceability
- Each bottle is supplied as a complete system, with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Caratteristiche e vantaggi
- Easier and safer handling with the award winning design. Specially shaped gripping zones on both sides of the bottle
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands
- Significantly faster opening or closing with the shortened bottle thread which remains fully compatible with GL 45 closures and other accessoires
- The pre-defined labelling areas are compatible with the dedicated DURAN® self-adhesive YOUTILITY bottle labels for unambigious bottle identification
- Space-saving design. The slimmer bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
- The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Award winning product design. Winner of seven international awards: Red Dot 2014, iF 2014, Silver IDEA International Excellence Award 2014, GOOD DESIGN AWARD Japan 2014, Special Mention German Design award 2015, Nominert DESIGNPREIS DER BUNDESREPUBLIC DEUTSCHLAND 2015, Silver DDC "Good Design 15" Award.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742010-4EA | 4061832802268 |