Documenti fondamentali
Z689947
Sigma-Aldrich
Aldrich® column clamp
size S
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
Misura
S
L × diam.
211 mm × 11 mm , arm
Capacità ganasce
65-89 mm
Categorie correlate
Descrizione generale
Corrosion resistant stainless steel. Ideal for holding chromatography columns and large cylindrical glassware. A large, flat thumbscrew/worm drive permits maximum tightening or removal in minimum time.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)