Z687707
Replacement NMR needle with bevel tip for eVol® electronic syringe
for 5 μL, L × O.D. 11.5 cm × 0.5 mm, gauge size 25
About This Item
Codice UNSPSC:
41121705
NACRES:
SB.32
Prodotti consigliati
Gauge
25
Produttore/marchio commerciale
SGE 036920
Lungh. × D.E.
11.5 cm × 0.5 mm
Categorie correlate
Descrizione generale
The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The beveled tip replacement needles come in multiple length and gauge options to fit your specific needs.
Note legali
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z687707-2EA | 4061833694572 |