Z687286
Primary HEPA filter with vapor after-filter module for Aldrich® compact ductless air cleaning system
Includes 6 in. deep HEPA filter with 2 in. deep refillable adsorption module
About This Item
Codice UNSPSC:
41104919
NACRES:
NB.43
Prodotti consigliati
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z687286-1EA | 4061832965420 |