Documenti fondamentali
Z679119
Anti-static unisex laboratory coats
size S
Sinonimo/i:
laboratory apparel
About This Item
Codice UNSPSC:
46181532
NACRES:
NB.35
Misura
S
Caratteristiche
CE compliant
Disponibilità
not available in USA and Canada
Descrizione generale
Tough white 210 g/m cotton/polyester reusable laboratory coats, full-length with a Nega-stat grid offering electrostatic protection.
  • Concealed stud-fastening front
  • Two hip pockets and one breast pocket, all reinforced with bar tacks
  • Finished length 100 cm, with center back vent
Conforms to prEN1149-3:2001
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)