Documenti fondamentali
Z678643
2mag MIX 6 multi position stirrer
AC/DC input 100 / 240 V AC, US 2-pin plug
Sinonimo/i:
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Materiali
stainless steel plate
Caratteristiche
positions 6
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
2mag 30006
Parametri
100-2000 rpm speed
Dimensioni della piastra × Largh. × Lungh.
× 245 mm × 400 mm
Volume max.
1-3000 mL , per stirring position
Descrizione generale
Ultra-flat magnetic stirrer with 6 stirring positions, up to 6,000 mL stirring volume, suitable for 6 x 1,000 mL flasks.
Powerful 6 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept, extremely wide speed range of 100 to 2,000 rpm, 100% synchronized speed, jerk-free stirring even at low speeds, 4-step power settings of the stirrer, high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)