Z674389
Duran® GLS 80 caps and accessories
GL 18 blanking cap, red
Sinonimo/i:
1170682
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Produttore/marchio commerciale
DURAN® 1170682
Categorie correlate
Descrizione generale
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Caratteristiche e vantaggi
- Manufactured from PP, PTFE, EPDM
- The flexible modular system allows an individual combination of the different components according to requirements and application
- Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts
- Sterile pressure equalisation is possible by using the syringe filter
- Unused ports can be sealed off with a blank GL 18 screw cap
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z674389-2EA | 4061832801001 |