Z616893
Vacuum accessories for Buchi® vacuum pumps
Woulff flask
About This Item
Codice UNSPSC:
41105101
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Büchi 047170
Descrizione generale
This optional Woulff flask may be connected in front of pump as a vacuum reservoir to protect pump from liquids accidentally sucked into vacuum line. Recommended in stand-alone systems using a V-700/710 pump. Includes retaining ring for V-700.
Note legali
Büchi is a registered trademark of Büchi Labortechnik AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z616893-1EA | 4061832773735 |