Z615684
Julabo® bath test tube racks
to fit 90 test tubes, for water baths, diam. 12-13 mm, polypropylene
Sinonimo/i:
tube holder
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.77
Prodotti consigliati
Materiali
polypropylene
Caratteristiche
Holds 90 x 12-13 mm tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Julabo 8970381
Parametri
+80 °C temp. limit
Diametro
12-13 mm
Categorie correlate
Descrizione generale
Polypropylene tray and test tube inserts for use up to +80 °C. When tempering solvent-containing or acidic samples above 60 °C, it is recommended that stainless steel test tube inserts are used, as the plastic can be attacked.
Note legali
Julabo is a registered trademark of Julabo USA, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)