Z556343
Aldrich® Sure/Stor™ adapter
joint: ST/NS 14/20
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: ST/NS14/20
Descrizione
with PTFE J Young valve and SafetyBarb®
hose connector
Categorie correlate
Descrizione generale
Converts round-bottom flasks to Sure/Stor flasks.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC
Ricambio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)