Z551031
Graduated Valumetric™ bottles
clear soda-lime glass bottle, capacity 250 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 24 ea
Sinonimo/i:
W216974
About This Item
Prodotti consigliati
Materiali
(PTFE faced LDPE liner)
clear soda-lime glass bottle
round bottle
white polypropylene cap
Giunto
Joints threaded neck (58-400)
Caratteristiche
with cap
Confezionamento
case of 24 ea
Produttore/marchio commerciale
WHEATON® W216974
Capacità
250 mL
diam. × alt.
62 mm × 127 mm
Categorie correlate
Descrizione generale
Caratteristiche e vantaggi
- Clear, USP Type III soda-lime glass
- Graduated in mL and ounces
- Easy removal of contents
- Available with caps attached or bulk packs without caps
- HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
- Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z551031-1PAK | 4061832831206 |