Z502200
Glindemann PTFE sealing rings
for 19/22, 19/38 joints
Sinonimo/i:
ptfe ring, ptfe sealing
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Prodotti consigliati
Giunto
joint 19/22
joint 19/38
Confezionamento
pkg of 50 ea
Produttore/marchio commerciale
Glindemann K19
Campo di misura della temperatura
≤250 °C
Descrizione generale
- This narrow ring seal is an alternative to a PTFE joint sleeve
- Greaseless protection for air- and moisture- sensitive compounds
- Can be used for prolonged storage under vacuum
- Slide easily onto medium and full length taper joints
- Can be used with Keck® clip and similar metal joint clips
Note legali
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z502200-1PAK | 4061833328156 |