Documenti fondamentali
Z420204
Sigma-Aldrich
Flaskup® holders assortment pack
3 ea of 10, 25, 50, and 100 mL sizes
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Materiali
polypropylene
Sterilità
non-sterile
Confezionamento
set of 1
Produttore/marchio commerciale
Bel-Art F38951-2008
Descrizione generale
Flaskup®olypropylene flask holders, assorted pack of 12. These unique holders are perfect for keeping round and tapered bottom flasks upright. Inside wall has a slight taper to the open bottom; allows a variety of flask shapes and sizes to fit into one holder .Wide base keeps flasks stable, side slots can be used as sight windows.
Caratteristiche e vantaggi
  • Molded from polypropylene plastic
  • Durable and easy to clean
  • Autoclavable at 121ºC (250ºF)
Note legali
Flaskup is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)