Documenti fondamentali
Z414379
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock on side-arm
joint: ST/NS 24/40, PTFE stopcock
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Materiali
PTFE stopcock
Giunto
joint: ST/NS24/40
Diam. ingresso
14 mm
Descrizione generale
Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)