Z405574
J. Young high-vacuum PTFE valve, straight through
stopcock size 8 mm, side arm O.D. 11 mm
About This Item
Codice UNSPSC:
40141600
NACRES:
NB.79
Prodotti consigliati
Produttore/marchio commerciale
J. Young
D.E. braccio laterale
11 mm
Misura rubinetto
8 mm
Descrizione generale
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
Features:
- PTFE piston with unique PTFE-sheathed elastomer O-ring seals
- Grease-free glass-to-PTFE seals
- High-vacuum to 10-6 torr
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z405574-1EA | 4061826668740 |