Z305359
Bransonic® Model B200 cleaner
AC/DC input 240 V AC
Sinonimo/i:
ultrasonic bath sonicator, ultrasonic cleaning bath
About This Item
Codice UNSPSC:
41103706
NACRES:
NB.74
Prodotti consigliati
Caratteristiche
CE compliant
CSA approved
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Branson 100-951-011 (NA)000-001-100 (Europe)
Descrizione generale
Low-cost, portable, ultrasonic cleaner has the same cleaning ability as larger models. SS tank with Noryl® base, parts basket, and plastic cover. Cleaner automatically shuts off after 5 min.
Note legali
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Noryl is a registered trademark of Sabic Innovative Plastics IP B.V.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z305359-1EA | 4061833295083 |
Z305359EU-1EA | 4061833295090 |
Z305359GB-1EA |