Documenti fondamentali
Z303151
Quickfit® Liebig condenser
L 160 mm, joint: ST/NS 14/23
Sinonimo/i:
C1/11/SC
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Giunto
joint: ST/NS14/23
Produttore/marchio commerciale
Quickfit C1/11/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Lunghezza
160 mm
Area superficiale
60 cm2
Descrizione generale
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)