Documenti fondamentali
Z278750
Sigma-Aldrich
Bytac® surface protection laminate
size × thickness 12 1/2 in. × 15 ft × 0.254 mm
Sinonimo/i:
bench protection, benchtop laminate, worksurface protection
About This Item
Codice UNSPSC:
41122300
NACRES:
NB.35
Materiali
vinyl backing
Confezionamento
pack of 1 ea
Misura × spessore
12 1/2 in. × 15 ft × 0.254 mm
Descrizione generale
Add the benefits of FEP to any flat surface. Bytac protects surfaces from absorption of toxic and radioactive materials. The laminate consists of FEP resin supported on a backing of vinyl or aluminum. The backing has a pressure sensitive acrylic adhesive that adheres to most surfaces. Vinyl backing is recommended for applications up to 200 °F (93 °C). Aluminum backing is for high-temperature applications and can be used up to 360 °F (182 °C).
  • non-stick; easy to clean
  • chemically inert
Note legali
Bytac is a registered trademark of Saint-Gobain Performance Plastics Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Whatman® Benchkote® Benchkote surface protector, sheets, 460 x 570 mm, 100/pk
WHA2300917
Whatman® Benchkote®
Whatman® Benchkote® Benchkote surface protector, reel, 460 mm x 50 m, 1/pk
WHA2300731
Whatman® Benchkote®
Whatman® Benchkote® Benchkote surface protector, reel, 920 mm x 50 m, 1/pk
WHA2300772
Whatman® Benchkote®