Z268828
Safe-Lab™ Thermometer/Tubing Adapter
joint: ST/NS 24/40, opening size 6-7 mm
Sinonimo/i:
thermometer holder
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Giunto
joint: ST/NS24/40
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F20309-1716
Dimens. apertura
6-7 mm
Descrizione generale
PTFE body with Viton® O-ring and PP screw-cap with inside Viton O-ring for air-tight work. Loosen cap for depth adjustment of thermometer or tubing.
Note legali
Safe-Lab is a trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z268828-1EA | 4061832257891 |