Documenti fondamentali
Z261149
Sigma-Aldrich
Nalgene® PassPort IP2 bottles
Wide-mouth, capacity 250 mL
Sinonimo/i:
nalgene bottle, nalgene bottles, nalgene hdpe
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
high-density polyethylene
polypropylene cap
round translucent bottle
Giunto
Joints threaded neck (43-415)
Caratteristiche
with cap
Confezionamento
pack of 12 (bottles)
Produttore/marchio commerciale
Nalgene® 2199-0008
Diametro bottiglia × alt.
61 mm × 131 mm
Capacità
250 mL
Descrizione generale
Bottles suitable for shipping. These bottles are in compliance with the new United Nations Performance-Oriented Packaging regulations for shipping. IP2 marking, material, volume, and registration notch molded into bottom of bottle. Leakproof. Evaluated at 15 psi.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PassPort is a trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)