Z261025
Nalgene® polypropylene copolymer separatory funnel
capacity 250 mL
Sinonimo/i:
liquid separation funnel, plastic separation device
About This Item
Codice UNSPSC:
41121809
eCl@ss:
32035600
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene copolymer bottle (PPCO)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Nalgene 4300-0250
Capacità
250 mL
Dimens. tappo
33 mm
Categorie correlate
Descrizione generale
Break-resistant and nearly transparent. Phase interface visible down to stopcock. PTFE stopcock and Tefzel® screw closure are leakproof.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z261025-1EA | 4061832616452 |