Z234230
Bend-A-Safety shield
H × W 24 in. × 48 in.
About This Item
Codice UNSPSC:
46181702
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
splash goggles included
Alt. × largh.
24 in. × 48 in.
Descrizione generale
Flexible benchtop safety workshield. Constructed of 19 gauge 2x2 steel mesh core which fits into a translucent PVC film cover with a zip fastener along one edge. Freestanding; this shield can be formed into a variety of shapes. The mesh holds the shape while the vinyl cover provides protection.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z234230-1EA | 4061832331997 |