Z231312
Giant-size molecular model kit
basic kit
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
Prodotti consigliati
Descrizione
no. of flexible bonds 16
no. of rigid bonds 50
Caratteristiche
no. of atoms 32
Descrizione generale
Large, lightweight molecular models for classroom demonstration of organic and inorganic structures. For building models to scale of 20 cm/Å and 8 cm/Å. Bond lengths range from 5.5 to 27 cm. Atomic centers are 5 cm in diameter, finished in standard atom colors, and have a reinforced vinyl receptor at each bond position for insertion of connecting rods (bonds). All model kits include storage box and instructions. Additional atoms and bonds are available separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z231312-1KT | 4061833577622 |