Documenti fondamentali
Z225614
Stainless steel forceps
Specimen, L 10 in.
About This Item
Codice UNSPSC:
41122405
NACRES:
NB.35
Confezionamento
pack of 1 ea
Lunghezza
10 in.
Descrizione generale
with serrated tips
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Stainless steel forceps Dumont
Z168742
Stainless steel forceps
Bayonet forceps stainless steel
F2394
Bayonet forceps
Dressing tissue forceps L 6 in.
F4267
Dressing tissue forceps
Dressing tissue forceps L 8 in.
F4392
Dressing tissue forceps
Stainless steel forceps Fine tip, straight
Z168777
Stainless steel forceps
Stainless steel forceps Fine tip, curved
Z168785
Stainless steel forceps
Stainless steel forceps Sharp-point
Z168718
Stainless steel forceps
Stainless steel forceps Reverse action
Z168769
Stainless steel forceps
Tweezers Style #4, needle-sharp, anti-magnetic stainless steel
T5790
Tweezers
Global Trade Item Number
SKUGTIN
Z225614-1EA4061833299487