Z225029
Aldrich® check-valve bubbler
T-inlet tube, side outlet
Sinonimo/i:
bubblers, scientific bubblers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Capacità
15 mL
Categorie correlate
Descrizione generale
Filling
Position the bubbler on it′s side with the left hose barb (as shown in the drawing) pointing straight up, then pipette 15 mL of oil through the hose barb into the bubbler reservoir. Position bubbler vertically as shown in drawing and it is ready to use.
Principal of operation
Gas enters the bubbler from the hose barb on the right and flows through the bubbler, exiting at the left. The gas pressure unseats the check-valve ball to release the pressure, bubbling up through the oil and out the left side. The check-valve ball reseats itself when pressure is neutralized. The advantage of a check-valve over bubbler such as Z223719 which does not have one, is that it prevents oil from being drawn from the bubbler reservoir into the reaction if a sudden negative pressure occurs.
Position the bubbler on it′s side with the left hose barb (as shown in the drawing) pointing straight up, then pipette 15 mL of oil through the hose barb into the bubbler reservoir. Position bubbler vertically as shown in drawing and it is ready to use.
Principal of operation
Gas enters the bubbler from the hose barb on the right and flows through the bubbler, exiting at the left. The gas pressure unseats the check-valve ball to release the pressure, bubbling up through the oil and out the left side. The check-valve ball reseats itself when pressure is neutralized. The advantage of a check-valve over bubbler such as Z223719 which does not have one, is that it prevents oil from being drawn from the bubbler reservoir into the reaction if a sudden negative pressure occurs.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z225029-1EA | 4061837617553 |