Z191566
Aldrich® hose clamp kits
115 pieces in assorted sizes from 11/32 to 15/8 in. o.d.
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
Prodotti consigliati
Descrizione generale
Non-corroding, lightweight nylon clamps secure Chem-Flex™ and standard laboratory tubing to glassware, hose fittings, and transfer needles. Provides effective hose-to-fitting seal by squeezing clamp jaws with pliers, lateral pressure on jaws will release. Usable temperature range: −25 to 225 °F. Pressure to 150 psi.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z191566-1KT | 4061833645512 |