Documenti
Z189855
Sigma-Aldrich
Spinplus® magnetic stirring bar
size 3/4 in. × 3/4 in.
Sinonimo/i:
Stir Bar, Stirbar, Stirring Bar
About This Item
Codice UNSPSC:
41103806
eCl@ss:
32100100
NACRES:
NB.72
Materiali
PTFE-coated
cross PTFE-coated bar
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F37144-0034
Lunghezza
19.1 mm
Diametro
9.5 mm
Misura
3/4 in. × 3/4 in.
Descrizione generale
Cross-shaped bars create a deep vortex for stable operations.
Note legali
Spinplus is a registered trademark of SP Scienceware, a division of SP