Documenti fondamentali
Z185159
Sigma-Aldrich
Aluminum foil
W × L 18 in. × 500 ft, thickness 0.001 in.
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.35
Sterilità
non-sterile
Produttore/marchio commerciale
Sigma-Aldrich
Largh. × lungh.
18 in. × 500 ft
Spessore
0.001 in.
applicazioni
PFAS testing
Descrizione generale
Easily covers laboratory dishes and forms inexpensive liners for bench tops, drawers, and trays. Easy cleanup in case of spill or contamination.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Aluminum foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
326852
Aluminum
Aluminum foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
326860
Aluminum
Aluminum IRMM®, certified reference material, 0.1 mm foil
IRMM523A
Aluminum
Aluminum foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
266957
Aluminum
Aluminum foil, thickness 0.10 mm, size 50 × 50 mm, purity 99.5%
GF00644554
Aluminum
Aluminum foil, thickness 0.05 mm, length 5 m, purity 99%
GF08708520
Aluminum
AlumaSeal® 384 film size 38 μm, thick aluminum foil sealing film for use with 384 well plates, non-sterile
Z721557
AlumaSeal® 384 film