Documenti fondamentali
Z178012
Sigma-Aldrich
Scienceware® POXYGRID® test-tube rack
Holds 108 x 15-16 mm tubes
Sinonimo/i:
tube holder
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.35
Materiali
epoxy (coated steel)
Caratteristiche
Holds 108 x 15-16 mm tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F18759-0001
Lungh. × Largh. × Alt.
237 mm × 186 mm × 67 mm
Descrizione generale
Welded steel wire is coated with heat-cured, thermosetting epoxy to protect from rust and corrosion. The coating is unaffected by most organic solvents, weak acids and alkalis, and has excellent abrasion resistance and adhesion to steel substrate.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Note legali
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)