Documenti fondamentali
Z177857
Sigma-Aldrich
Scienceware® Ellipso-Spoon® sampler
L 18 cm
Sinonimo/i:
curved laboratory spatula, laboratory spoon
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Produttore/marchio commerciale
Bel-Art H368060018
Lunghezza
18 cm
Descrizione generale
Special curved shape permits contact with all corners of vessels, bottles, or containers. No corner out of reach and no loss of materials. Handles crystals, powders, granulated and solid substances, even thick liquids or delicate substances. SS, will not bend. Has both spoon end and spatula end.
Note legali
Ellipso-Spoon is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Lab spoons stainless steel, L 7 1/2 in.
Sigma-Aldrich
Z511455
Lab spoons
Micro spatulas one scraper/spoon end, L 6 1/2 in. (165 mm)
Z193216
Micro spatulas
Global Trade Item Number
SKUGTIN
Z177857-1EA4061832257280